Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-FADD (S194) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7047
This item is not returnable.
View return policy
Description
FADD Monoclonal specifically detects FADD in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
FADD (p ser194) | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1, MORT1Fas-associating protein with death domain, protein FADD | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8772 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Immunocytochemistry | |
709736 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FADD | |
Phosphopeptide containing the human FADD S194 site | |
100 μg | |
Primary | |
Detects human FADD when phosphorylated on Ser194. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction