Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-GSK-3 beta (S9) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $526.00
Specifications
Antigen | GSK-3 beta |
---|---|
Clone | 1253C |
Dilution | Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Description
GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
GSK-3 beta | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK3 beta, GSK-3 beta, GSK3B, GSK3beta isoform | |
GSK3B | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1253C | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P49840 | |
2932 | |
Phosphopeptide containing the human GSK-3 beta S9 site | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human GSK-3 beta when phosphorylated at S9 in Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title