Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-LAT (Y161) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6334
This item is not returnable.
View return policy
Description
LAT Monoclonal specifically detects LAT in Human samples. It is validated for Western Blot.Specifications
| LAT (p tyr161) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| O43561 | |
| LAT | |
| Phosphopeptide containing the human LAT Y161 site Accession # O43561 | |
| 100 μg | |
| Primary | |
| Detects human Phospho-LAT (Y161) in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 627945 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27040 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction