Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-LRP-6 (S1490) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF6649
This item is not returnable.
View return policy
Description
LRP-6 Polyclonal specifically detects LRP-6 in Human samples. It is validated for Western Blot.Specifications
LRP-6 (p ser1490) | |
Polyclonal | |
Unconjugated | |
ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP6, LRP-6 | |
Rabbit | |
Affinity Purified | |
RUO | |
4040 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
LRP6 | |
Phosphopeptide containing the human LRP-6 S1490 site. | |
100 μg | |
Primary | |
Detects human LRP-6 when phosphorylated at S1490 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction