Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-MEK1 (T386) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems PPS076
This item is not returnable.
View return policy
Description
MEK1 Polyclonal specifically detects MEK1 in Human samples. It is validated for Western Blot.Specifications
MEK1 (p thr386) | |
Polyclonal | |
Western Blot 1:1000 dilution | |
MAP2K1 | |
Phosphopeptide corresponding to amino acid residues surrounding the human phospho-T386 of MEK1 | |
100 μL | |
Primary | |
Human ∼45 kDa MEK1 phosphorylated at T386 | |
For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year. |
Western Blot | |
Unconjugated | |
dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAP2K1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1) | |
Rabbit | |
Affinity Purified | |
RUO | |
5604 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction