Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-MSK1(S376)/MSK2(S360) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB25181SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB25181SP 25 μg
MAB25181 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB25181SP Supplier R&D Systems Supplier No. MAB25181SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MSK1/MSK2 Monoclonal specifically detects MSK1/MSK2 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen MSK1/MSK2
Applications Immunocytochemistry
Classification Monoclonal
Clone 1013F
Conjugate Unconjugated
Dilution Immunocytochemistry 1-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. O75582
Gene Alias 1110069D02Rik, 3110005L17Rik, 6330404E13Rik, RPS6KA4, RPS6KA5, RSKB
Host Species Rabbit
Immunogen Phosphopeptide containing the human MSK1(S376) site. Accession # O75582
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human MSK1 and MSK2 when phosphorylated at S376 and S360, respectively.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.