Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Phospho-MSPR/Ron DuoSet IC ELISA
SDP

Catalog No. DYC19475 Shop All Novus Biologicals Products
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC19475 5 Plates
DYC19472 2 Plates
DYC1947E 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC19475 Supplier R&D Systems™ Supplier No. DYC19475

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

The Human Phospho-MSP R/Ron DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human MSPR/Ron.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
For Use With (Application) ELISA
Gene ID (Entrez) 4486
Product Line R&D Systems
Synonym CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4
Packaging Quantity 5 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Signaling
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.