Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-NFATC1 (S172) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5640SP
This item is not returnable.
View return policy
Description
NFATC1/NFAT2 Monoclonal specifically detects NFATC1/NFAT2 in Human samples. It is validated for Western Blot.Specifications
NFATC1/NFAT2 (p ser172) | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
MGC138448, NFAT transcription complex cytosolic component, NFAT2, NFAT2cytoplasmic 1, NFATC, NF-ATC, NFATc1, NF-ATc1, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4772 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot | |
679340 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
NFATC1 | |
Phosphopeptide containing the human S172 site. | |
25 μg | |
Primary | |
Detects human NFATC1 in direct ELISAs. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction