Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-p38 alpha (T180/Y182) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8692SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8692SP 25 μg
MAB8692 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8692SP Supplier R&D Systems Supplier No. MAB8692SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

p38 alpha Monoclonal specifically detects p38 alpha in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen p38 alpha
Applications Immunocytochemistry
Classification Monoclonal
Clone 948243
Conjugate Unconjugated
Dilution Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q16539
Gene Symbols MAPK14
Host Species Rat
Immunogen Phosphopeptide containing human p38 alpha T180/Y182 site Accession # Q16539
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1432
Test Specificity Detects human p38 alpha when phosphorylated at T180/Y182.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.