Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-p53 (S37) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF3306
This item is not returnable.
View return policy
Description
p53 Polyclonal specifically detects p53 in Human samples. It is validated for Western Blot.Specifications
p53 (p ser37) | |
Polyclonal | |
Unconjugated | |
Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53 | |
Rabbit | |
Affinity Purified | |
RUO | |
7157 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.4 ug/mL | |
TP53 | |
Phosphopeptide containing human p53 S37 site | |
50 μg | |
Primary | |
Detects human p53 when phosphorylated at S37. Does not recognize p53 unphosphorylated at S37. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction