Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-p53 (S392) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2996100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2996100 100 μg
MAB2996SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2996100 Supplier R&D Systems Supplier No. MAB2996100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

p53 Monoclonal specifically detects p53 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen p53
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2185A
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P04637
Gene Alias Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53
Host Species Rabbit
Immunogen Phosphopeptide containng the human p53 S392 site Accession # P04637
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7157
Test Specificity Detects human p53 when phoshorylated at S392
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.