Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-p53 (S46) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB1489
This item is not returnable.
View return policy
Description
p53 Monoclonal specifically detects p53 in Human samples. It is validated for Western Blot.Specifications
p53 (p Ser46) | |
Monoclonal | |
Unconjugated | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
TP53 | |
Phosphopeptide containing the human p53 S46 site | |
100 μg | |
Primary | |
Detects human phospho-p53 (S46) in Western blots. | |
12 months from date of receipt, -20 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 degreesC under sterile conditions after opening. | |
IgG |
Western Blot | |
1022G | |
Western Blot 1:1000 dilution | |
Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
7157 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Contains <0.1% Sodium Azide, which is not hazardous at this concentration according to GHS classifications. Refer to SDS for additional information and handling instructions.
Spot an opportunity for improvement?Share a Content Correction