Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-p70 S6 Kinase (T229) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8964
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8964 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8964 Supplier R&D Systems Supplier No. MAB8964
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

p70 S6 Kinase/S6K Monoclonal specifically detects p70 S6 Kinase/S6K in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen p70 S6 Kinase/S6K
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1015B
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunocytochemistry 1:200 dilution
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, RPS6KB1, S6K, S6K1, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A
Gene Symbols RPS6KB1
Host Species Rabbit
Immunogen Phosphopeptide containing human p70S6K T229
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6198
Test Specificity Detects human p70 S6 Kinase when phosphorylated at T229.
Target Species Human
Content And Storage 12 months from date of receipt, -20 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 degreesC under sterile conditions after opening.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.