Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-PLC-gamma 1 (Y783) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7454
This item is not returnable.
View return policy
Description
PLC-gamma 1 Monoclonal specifically detects PLC-gamma 1 in Human samples. It is validated for Western Blot.Specifications
PLC-gamma 1 (p tyr783) | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
EC 3.1.4.11, gamma 1 (formerly subtype 148), monophosphatidylinositol phosphodiesterase, NCKAP3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1, phosphatidylinositol phospholipase C, phosphoinositidase C, phosphoinositide phospholipase C, Phosphoinositide phospholipase C-gamma-1, phospholipase C, gamma 1, phospholipase C-148, Phospholipase C-gamma-1, Phospholipase C-II, PLC11-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma 1, PLC148, PLC-148, PLCG1, PLCgamma 1, PLC-gamma-1, PLC-II1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, PPLCA, triphosphoinositide phosphodiesterase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5335 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot | |
764518 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PLCG1 | |
Phosphopeptide containing the human PLC-gamma 1 Y783 site | |
100 μg | |
Primary | |
Detects human Phospho-PLC-gamma 1 (Y783) in direct ELISAs. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction