Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-Progesterone R/NR3C3 (S294) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF5955SP
This item is not returnable.
View return policy
Description
Progesterone R/NR3C3 Polyclonal specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Western Blot.Specifications
Progesterone R/NR3C3 (p ser294) | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
NR3C3, NR3C3PRNuclear receptor subfamily 3 group C member 3, PGR, progesterone receptor, ProgesteroneR | |
Rabbit | |
Affinity Purified | |
RUO | |
5241 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P06401 | |
PGR | |
Phosphopeptide containing human Progesterone RB S294 site Accession # P06401 | |
25 μg | |
Primary | |
Detects human Phospho-Progesterone R/NR3C3 (S294) in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction