Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-PYK2/FAK2 (Y402) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6210
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6210 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6210 Supplier R&D Systems Supplier No. MAB6210
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PYK2/FAK2 Monoclonal specifically detects PYK2/FAK2 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen PYK2/FAK2 (p Tyr402)
Applications Western Blot
Classification Monoclonal
Clone 592918
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q14289
Gene Alias CADTK, CADTKCAK-beta, CAK beta, CAKB, Calcium-dependent tyrosine kinase, Cell adhesion kinase beta, EC 2.7.10, FADK2, FAK2, FAK2EC 2.7.10.2, Focal adhesion kinase 2, FRNK, PKB, ppyk2, Proline-rich tyrosine kinase 2, protein kinase B, protein tyrosine kinase 2 beta, protein-tyrosine kinase 2-beta, PTK, PTK2B, PTK2B protein tyrosine kinase 2 beta, PYK2Related adhesion focal tyrosine kinase, RAFTK, RAFTKFADK 2
Gene Symbols PTK2B
Host Species Mouse
Immunogen Phosphopeptide containing the human PYK2/FAK2 Y402 site Accession # Q14289
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2185
Test Specificity Detects human PYK2/FAK2 when phosphorylated at Y402.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.