Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-Ret (Y905) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF3269
This item is not returnable.
View return policy
Description
Ret Polyclonal specifically detects Ret in Human samples. It is validated for Western Blot.Specifications
Ret (p tyr905) | |
Polyclonal | |
Unconjugated | |
CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
Rabbit | |
Affinity Purified | |
RUO | |
5979 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
RET | |
Phosphopeptide containing human Ret Y905 site | |
50 μg | |
Primary | |
Detects human Ret when phosphorylated at Y905 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction