Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-Ribosomal Protein S6/RPS6 (S235/S236) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB54361SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB54361SP 25 μg
MAB54361100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB54361SP Supplier R&D Systems Supplier No. MAB54361SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Ribosomal Protein S6/RPS6 Monoclonal specifically detects Ribosomal Protein S6/RPS6 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Ribosomal Protein S6/RPS6
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 973210
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P62753
Gene Alias Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, RPS6, S6
Host Species Rat
Immunogen Phosphopeptide containing human Ribosomal Protein S6 S235/S236 sites Accession # P62753
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 6194
Test Specificity Detects human Ribosomal Protein S6 when dually phosphorylated at S235 and S236.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.