Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-SLP-76/LCP2 (Y145) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7474
This item is not returnable.
View return policy
Description
SLP-76/LCP2 Monoclonal specifically detects SLP-76/LCP2 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
SLP-76/LCP2 (p Tyr145) | |
Monoclonal | |
LYOPH | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q13094 | |
LCP2 | |
Phosphopeptide containing the human SLP-76/LCP2 Y145 site Accession # Q13094 | |
100 μg | |
Primary | |
Detects human Phospho-SLP-76/LCP2 in direct ELISAs. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
776503 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LCP2, lymphocyte cytosolic protein 2, lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of76kDa), lymphocyte cytosolic protein 2 (SH2 domain-containing leukocyte protein of76kD), SH2 domain-containing leukocyte protein of 76 kDa, SH2 domain-containing leukocyte protein of 76kD, SLP76, SLP-76, SLP-76 tyrosine phosphoprotein, SLP7676 kDa tyrosine phosphoprotein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3937 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction