Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-STAT1 (Y701) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2894
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2894 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2894 Supplier R&D Systems Supplier No. MAB2894
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

STAT1 Monoclonal specifically detects STAT1 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry.

Specifications

Antigen STAT1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 1086B
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. NP009330
Gene Alias CANDF7, DKFZp686B04100, ISGF-3, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1, STAT91
Gene Symbols STAT1
Host Species Rabbit
Immunogen Phosphopeptide containing the human STAT1 Y701 site Accession # NP009330
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6772
Test Specificity Detects human STAT1 when phosphorylated at Y701 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.