Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-STAT5a/b (Y694/Y699) Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

$214.00 - $538.00

Specifications

Antigen STAT5a/b
Clone 1247C
Dilution Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Applications Western Blot, Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB41901SP
GREENER_CHOICE
View Documents
R&D Systems
MAB41901SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB41901
GREENER_CHOICE
View Documents
R&D Systems
MAB41901
100 μg
Each for $538.00
Only null left
Add to Cart
 
Description

Description

STAT5a/b Monoclonal specifically detects STAT5a/b in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications

Specifications

STAT5a/b
Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
MGF, STAT5, STAT5A signal transducer and activator of transcription 5A
Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694) Accession # P51692
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human STAT5a/b when phosphorylated at Y964/Y699 in direct ELISAs and Western blots.
1247C
Western Blot, Flow Cytometry
Unconjugated
Rabbit
Human
P51692
6776
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.