Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-STAT5a/b (Y694/Y699) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4190SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4190SP 25 μg
MAB4190 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4190SP Supplier R&D Systems Supplier No. MAB4190SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

STAT5a/b Monoclonal specifically detects STAT5a/b in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen STAT5a/b
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 941212
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 0.3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P51692
Gene Alias MGF, STAT5, STAT5A signal transducer and activator of transcription 5A
Host Species Rat
Immunogen Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694). Accession # P51692
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6776
Test Specificity Detects human STAT5a/b when phosphorylated at Y694/Y699 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.