Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Phospho-VEGFR2/KDR DuoSet IC ELISA
SDP

Catalog No. DYC1766E Shop All R&D Systems Products
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC1766E 15 Plates
DYC17665 5 Plates
DYC17662 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC1766E Supplier R&D Systems™ Supplier No. DYC1766E
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Phospho-VEGFR2/KDR DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Phospho-VEGFR2/KDR DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human VEGFR2/KDR/Flk-1.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
For Use With (Application) ELISA
Gene ID (Entrez) 3791
Synonym CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2
Packaging Quantity 15 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.