Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-WNK1 (T60) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4720100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4720100 100 μg
MAB4720SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4720100 Supplier R&D Systems Supplier No. MAB4720100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

WNK1 Monoclonal specifically detects WNK1 in Human samples. It is validated for Western Blot.

Specifications

Antigen WNK1
Applications Western Blot
Classification Monoclonal
Clone 1259F
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. Q9H4A3
Gene Alias EC 2.7.11, EC 2.7.11.1, Erythrocyte 65 kDa protein, HSAN2hereditary sensory neuropathy, type II, HSN2MGC163339, hWNK1, KDP, KDPMGC163341, KIAA0344, Kinase deficient protein, p65, PHA2C, PRKWNK1, PRKWNK1PSK, prostate-derived sterile 20-like kinase, Protein kinase lysine-deficient 1, Protein kinase with no lysine 1, protein kinase, lysine deficient 1, serine/threonine-protein kinase WNK1, WNK lysine deficient protein kinase 1
Host Species Rabbit
Immunogen Phosphopeptide containing human WNK1 T60 site Accession # Q9H4A3
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65125
Test Specificity Detects human WNK1 when phosphorylated at T60 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.