Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-ZAP70 (Y493) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7694
This item is not returnable.
View return policy
Description
ZAP70 Monoclonal specifically detects ZAP70 in Human samples. It is validated for Western Blot.Specifications
ZAP70 (p tyr493) | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
70 kDa zeta-associated protein, EC 2.7.10, EC 2.7.10.2, mrtle, mur, SRK, SRKFLJ17679, STD, STDFLJ17670, Syk-related tyrosine kinase, tyrosine-protein kinase ZAP-70, TZK, ZAP-70, zeta-chain (TCR) associated protein kinase (70 kD), zeta-chain (TCR) associated protein kinase 70kDa, zeta-chain associated protein kinase, 70kD | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7535 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
797917 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ZAP70 | |
Phosphopeptide containing the human ZAP70 Y493 site | |
100 μg | |
Primary | |
Detects human ZAP70 when phosphorylated at Y493. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction