Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PI 3-Kinase p85 beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6777
This item is not returnable.
View return policy
Description
PI 3-Kinase p85 beta Monoclonal specifically detects PI 3-Kinase p85 beta in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| PI 3-Kinase p85 beta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| O00459 | |
| PIK3R2 | |
| E. coli-derived recombinant human PI 3-Kinase p85 beta Lys516-Gly642 Accession # O00459 | |
| 100 μg | |
| Primary | |
| Detects human PI 3-Kinase p85 beta in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) PI 3-Kinase p85 beta (aa 325-428), rhPI 3-Kinase p85 alpha , rhPI 3-Kinase p87, rhPIK3R4, or rhPIK3R5 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 572001 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| P85B, p85-beta, phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta, phosphatidylinositol 3-kinase regulatory subunit beta, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta), phosphoinositide-3-kinase, regulatory subunit 2 (beta), phosphoinositide-3-kinase, regulatory subunit 2 (p85 beta), phosphoinositide-3-kinase, regulatory subunit, polypeptide 2 (p85 beta), PI 3Kinase p85 beta, PI3K regulatory subunit beta, PI3-kinase regulatory subunit beta, PI3-kinase subunit p85-beta, PIK3R2, ptdIns-3-kinase regulatory subunit beta, ptdIns-3-kinase regulatory subunit p85-beta | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5296 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction