Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human pIgR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2717SP
This item is not returnable.
View return policy
Description
pIgR Monoclonal specifically detects pIgR in Human samples. It is validated for Western Blot, Simple Western.Specifications
pIgR | |
Monoclonal | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL | |
CAA51532 | |
PIGR | |
Mouse myeloma cell line NS0-derived recombinant human pIgR Lys19-Arg638 Accession # CAA51532 | |
25 μg | |
Primary | |
Detects human pIgR in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse pIgR is observed. | |
Human | |
Purified |
Western Blot | |
825724 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ22667, hepatocellular carcinoma associated protein TB6, Hepatocellular carcinoma-associated protein TB6, MGC125361, MGC125362, PIgR, Poly-Ig receptor, polymeric immunoglobulin receptor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5284 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction