Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PILR-alpha Alexa Fluor 488-conjugated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems FAB6484G
This item is not returnable.
View return policy
Description
PILR-alpha Polyclonal specifically detects PILR-alpha in Human samples. It is validated for Flow Cytometry.Specifications
PILR-alpha | |
Polyclonal | |
Flow Cytometry 5 uL/10^6 cells | |
Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha, PILRA, PILRalpha | |
Sheep | |
Affinity Purified | |
RUO | |
29992 | |
Human | |
IgG |
Flow Cytometry | |
Alexa Fluor 488 | |
Q9UKJ1 | |
PILRA | |
Mouse myeloma cell line NS0-derived recombinant human PILR-alpha Gln20-Thr196 Accession # Q9UKJ1 | |
100 Tests | |
Primary | |
Detects human PILR-alpha in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction