Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PILR-alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6484
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6484 Supplier R&D Systems Supplier No. MAB6484
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PILR-alpha Monoclonal specifically detects PILR-alpha in Human samples. It is validated for Western Blot.

Specifications

Antigen PILR-alpha
Applications Western Blot
Classification Monoclonal
Clone 462415
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UKJ1
Gene Alias Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha, PILRA, PILRalpha
Gene Symbols PILRA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PILR-alpha Gln20-Thr196 (predicted) Accession # Q9UKJ1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29992
Test Specificity Detects human PILR-alpha in direct ELISAs and Western blots. No cross-reactivity with recombinant human PILR-beta , recombinant mouse (rm) PILR-alpha , or rmPILR-beta is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.