Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PILR-alpha Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6484
This item is not returnable.
View return policy
Description
PILR-alpha Polyclonal specifically detects PILR-alpha in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
PILR-alpha | |
Polyclonal | |
Unconjugated | |
Q9UKJ1 | |
PILRA | |
Mouse myeloma cell line NS0-derived recombinant human PILR-alpha Gln20-Thr196 Accession # Q9UKJ1 | |
100 μg | |
Primary | |
Detects human PILR-alpha in direct ELISAs. | |
Human | |
IgG |
Flow Cytometry, CyTOF | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha, PILRA, PILRalpha | |
Sheep | |
Affinity Purified | |
RUO | |
29992 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction