Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PILR-beta Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4189
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4189 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4189 Supplier R&D Systems Supplier No. BAF4189
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PILR-beta Polyclonal specifically detects PILR-beta in Human samples. It is validated for Western Blot.

Specifications

Antigen PILR-beta
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9UKJ0
Gene Alias activating receptor PILRbeta, Activating receptor PILR-beta, Cell surface receptor FDFACT, cell surface receptor FDFACT1, cell surface receptor FDFACT2, FDFACT, FDFACT1, FDFACT2, paired immunoglobin-like receptor beta, paired immunoglobin-like type 2 receptor beta, paired immunoglobulin-like receptor beta, paired immunoglobulin-like type 2 receptor beta, PILRB, PILRbeta
Gene Symbols PILRB
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human PILR-beta isoform 1 Gln20-Ala189 Accession # Q9UKJ0
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29990
Test Specificity Detects human PILR-beta in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human PILR-alpha is observed and less than 5% cross-reactivity with recombinant mouse PILR-beta , rmPILR-alpha , and rmPILR-L is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.