Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PKC epsilon Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5134
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5134 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5134 Supplier R&D Systems Supplier No. MAB5134
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PKC epsilon Monoclonal specifically detects PKC epsilon in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen PKC epsilon
Applications Immunohistochemistry
Classification Monoclonal
Clone 666823
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q02156
Gene Alias EC 2.7.11, EC 2.7.11.13, MGC125656, nPKC-epsilon, PKCE, PKCEMGC125657, PRKCE, protein kinase C epsilon type, protein kinase C, epsilon
Gene Symbols PRKCE
Host Species Mouse
Immunogen E. coli-derived recombinant human PKC epsilon Gln580-Pro737 Accession # Q02156
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5581
Test Specificity Detects human PKC epsilon in direct ELISAs. In direct ELISAs, less than 20% cross-reactivity with recombinant human (rh) PKC delta and rhPKC iota is observed. No cross-reactivity with rhPKC alpha , beta 1, beta 2, gamma , eta , μ, nu , theta , or zeta is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.