Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PKC zeta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6666
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6666 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6666 Supplier R&D Systems Supplier No. MAB6666
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PKC zeta Monoclonal specifically detects PKC zeta in Human samples. It is validated for Western Blot.

Specifications

Antigen PKC zeta
Applications Western Blot
Classification Monoclonal
Clone 453513
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q05513
Gene Alias EC 2.7.11, EC 2.7.11.13, nPKC-zeta, PKC2, PKC2PKC-ZETA, PRKCZ, protein kinase C zeta type, protein kinase C, zeta
Gene Symbols PRKCZ
Host Species Mouse
Immunogen E. coli-derived recombinant human PKC zeta Ile454-Val592 Accession # Q05513
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5590
Test Specificity Detects human PKC zeta in direct ELISAs and Western blots. In direct ELISAs, less than 25% cross-reactivity with recombinant human (rh) PKC iota and rhPKC alpha and no cross-reactivity with rhPKC epsilon, eta, beta 1, beta 2, delta, gamma, nu, mu, or theta is observed. In Western blots, 100% cross-reactivity with recombinant human (rh) PKC iota and no cross-reactivity with rhPKC epsilon or eta is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.