Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PKM1/2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB72443SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB72443SP 25 μg
MB72443100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB72443SP Supplier R&D Systems Supplier No. MAB72443SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PKM2 Monoclonal specifically detects PKM2 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen PKM2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 945103
Conjugate Unconjugated
Dilution Western Blot 0.25-0.5 ug/mL, Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P14618
Gene Alias CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP3, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, p58, PK, muscle type, PK2, PK3, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK
Host Species Mouse
Immunogen E.coli-derived recombinant human PKM2, Ser434-Pro531, Accession No. P14618
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 5315
Test Specificity Detects human PKM2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.