Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PLA2G16 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6190
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6190 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6190 Supplier R&D Systems Supplier No. AF6190
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PLA2G16/HRASLS3 Polyclonal specifically detects PLA2G16/HRASLS3 in Human samples. It is validated for Western Blot.

Specifications

Antigen PLA2G16/HRASLS3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P53816
Gene Alias Adipose-specific phospholipase A2, AdPLA, AdPLAHRASLS3, EC 3.1.1.4, group XVI phospholipase A2, HRAS-like suppressor 3Ca-independent phospholipase A1/2, HRASLS3, HREV107, H-REV107-1, HREV107-3adipose-specific PLA2, HREV107H-rev 107 protein homolog, MGC118754, phospholipase A2, group XVI, Renal carcinoma antigen NY-REN-65
Gene Symbols PLA2G16
Host Species Sheep
Immunogen E. coli-derived recombinant human PLA2G16 Asp12-Asp132 Accession # P53816
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11145
Test Specificity Detects human PLA2G16 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.