Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PLA2G1B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5018
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5018 Supplier R&D Systems Supplier No. MAB5018
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PLA2G1B Monoclonal specifically detects PLA2G1B in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PLA2G1B
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 630823
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NM_000928
Gene Alias Group IB phospholipase A2, Phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2, phospholipase A2, group IB (pancreas), PLA2, PLA2A, PLA2AMGC119834, PLA2EC 3.1.1.4, PPLA2, PPLA2MGC119835
Gene Symbols PLA2G1B
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PLA2G1B Met1-Ser148 Accession # NM_000928
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5319
Test Specificity Detects human PLA2G1B in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) PLA2G2A, rhPLA2G7, recombinant mouse (rm) PLA2G2A, or rmPLA2R1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.