Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PLA2G7/PAF-AH/Lp-PLA2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5106100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5106100 100 μg
MAB5106SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5106100 Supplier R&D Systems Supplier No. MAB5106100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PLA2G7/PAF-AH/Lp-PLA2 Monoclonal specifically detects PLA2G7/PAF-AH/Lp-PLA2 in Human samples. It is validated for ELISA.

Specifications

Antigen PLA2G7/PAF-AH/Lp-PLA2
Applications ELISA
Classification Monoclonal
Clone 973010
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. Q13093
Gene Alias 2-acetyl-1-alkylglycerophosphocholine esterase, EC 3.1.1, EC 3.1.1.47,1-alkyl-2-acetylglycerophosphocholine esterase, Group-VIIA phospholipase A2, gVIIA-PLA2, LDL-associated phospholipase A2, LDL-PLA(2), LDL-PLA2, lipoprotein-associated phospholipase A2, LpPLA2, LP-PLA2, PAF acetylhydrolase, PAF-AH, PAFAHPAF 2-acylhydrolase, phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma), platelet-activating factor acetylhydrolase
Host Species Mouse
Immunogen E. coli-derived recombinant human PLA2G7/PAF-AH/Lp-PLA2 Phe22-Asn441 Accession # Q13093
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7941
Test Specificity Detects human PLA2G7Phe22-Asn441 in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.