Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Plasminogen Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1939SP
This item is not returnable.
View return policy
Description
Plasminogen Monoclonal specifically detects Plasminogen in Human samples. It is validated for Immunoprecipitation, Neutralization.Specifications
Plasminogen | |
Monoclonal | |
LYOPH | |
Immunoprecipitation 25 ug/mL, Neutralization 1 ug/mL | |
DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen, Plg | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5340 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Immunoprecipitation, Neutralization | |
270409 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PLG | |
Human plasma-derived Plasminogen Glu20-Asn810 | |
25 μg | |
Primary | |
Detects human PLG, human Plasmin, and the catalytic domain in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction