Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PLD1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5615SP
This item is not returnable.
View return policy
Description
PLD1 Polyclonal specifically detects PLD1 in Human samples. It is validated for Western Blot.Specifications
PLD1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Choline phosphatase 1, EC 3.1.4.4, hPLD1, Phosphatidylcholine-hydrolyzing phospholipase D1, phospholipase D1, phospholipase D1, phophatidylcholine-specific, phospholipase D1, phosphatidylcholine-specific, PLD 1 | |
Goat | |
Affinity Purified | |
RUO | |
5337 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q13393 | |
PLD1 | |
E. coli-derived recombinant human PLD1 Met1-Pro140 Accession # Q13393 | |
25 μg | |
Primary | |
Detects endogenous human PLD1 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction