Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PLD2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10123100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10123100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB10123100 Supplier R&D Systems Supplier No. MAB10123100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PLD2 Monoclonal specifically detects PLD2 in Human samples. It is validated for Western Blot.

Specifications

Antigen PLD2
Applications Western Blot
Classification Monoclonal
Clone 846224
Conjugate Unconjugated
Dilution Western Blot 2 μ/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O14939
Gene Alias Choline Phosphatase 2, HPLD2, PLD1C
Host Species Mouse
Immunogen E. coli-derived recombinant human PLD2, Met1-His117, Accession No. O14939
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 5338
Test Specificity Detects huma PLD2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.