Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PLD2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB10123SP
This item is not returnable.
View return policy
Description
PLD2 Monoclonal specifically detects PLD2 in Human samples. It is validated for Western Blot.Specifications
PLD2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Choline Phosphatase 2, HPLD2, PLD1C | |
E. coli-derived recombinant human PLD2, Met1-His117, Accession No. O14939 | |
25 μg | |
5338.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
846224 | |
Western Blot 2 ug/mL | |
O14939 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects huma PLD2 in direct ELISAs. | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction