Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pleiotrophin/PTN Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Pleiotrophin/PTN |
---|---|
Clone | 851407 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2521SP
![]() |
R&D Systems
MAB2521SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2521100
![]() |
R&D Systems
MAB2521100 |
100 μg |
Each for $502.50
|
|
|||||
Description
Pleiotrophin/PTN Monoclonal specifically detects Pleiotrophin/PTN in Human samples. It is validated for ELISA.Specifications
Pleiotrophin/PTN | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
P21246 | |
5764 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
851407 | |
ELISA | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
HARP, HBBM, HB-GAM, HBGF8, HBNF, HBNF1, HBNF-1, Heparin-binding brain mitogen, HNGF-8, NEGF1HBGF-8, neurite growth-promoting factor 1, neurite growth-promoting factor1), OSF-1, Osteoblast-specific factor 1, pleiotrophin, PTN | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Pleiotrophin/PTN Gly33-Asp168 Accession # P21246 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human Pleotrophin/PTN in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title