Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Plexin A4 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB58561G100UG
This item is not returnable.
View return policy
Description
Plexin A4 Monoclonal specifically detects Plexin A4 in Human samples. It is validated for Flow Cytometry.Specifications
Plexin A4 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9HCM2 | |
PLXNA4 | |
Chinese hamster ovary cell line CHO-derived recombinant human Plexin A4 Thr24-Pro1237 Accession # Q9HCM2 | |
100 μg | |
Primary | |
Detects human Plexin A4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Plexin A1, recombinant mouse (rm) Plexin A1 or rmPlexin A2 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Flow Cytometry | |
707206 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
A, DKFZp434G0625, FAYV2820, FLJ35026, FLJ38287, KIAA1550DKFZp566O0546, PLEXA4, plexin A4, plexin A4, B, plexin-A4, PLXNA4, PLXNA4A, PLXNA4B, PRO34003 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
91584 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction