Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Plexin B1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB37491V100UG
This item is not returnable.
View return policy
Description
Plexin B1 Monoclonal specifically detects Plexin B1 in Human samples. It is validated for Flow Cytometry.Specifications
Plexin B1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O43157 | |
PLXNB1 | |
NS0 mouse myeloma cell line transfected with human Plexin B1 Accession # O43157 | |
RUO | |
5364 | |
Human | |
IgG1 |
Flow Cytometry | |
559830 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
plexin B1, PLXN5, PLXNB1, SEP, Transmembrane Protein SEP | |
Mouse | |
100 μg | |
Primary | |
Detects human Plexin B1 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction