Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Plexin B2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5329
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5329 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5329 Supplier R&D Systems Supplier No. MAB5329
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Plexin B2 Monoclonal specifically detects Plexin B2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Plexin B2
Applications Western Blot
Classification Monoclonal
Clone 537237
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O15031
Gene Alias MM1, MM1KIAA0315dJ402G11.3, Nbla00445, PLEXB2, plexin B2, plexin-B2, PLXNB2
Gene Symbols PLXNB2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Plexin B2 Leu20-Arg1160 Accession # O15031
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23654
Test Specificity Detects human Plexin B2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Plexin A4, B1, B3, C1, D1, or recombinant mouse Plexin A1, A2, or A3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.