Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Plexin B3 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4958
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4958 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4958 Supplier R&D Systems Supplier No. BAF4958
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Plexin B3 Polyclonal specifically detects Plexin B3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Plexin B3
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Gene Alias PLEXB3, plexin B3, plexin-B3, PLEXR, PLXN6, PLXNB3
Gene Symbols PLXNB3
Host Species Sheep
Immunogen CHO cell-derived rhPlexin B3 extracellular domain
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5365
Test Specificity Detects human Plexin B3 in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse Plexin B3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.