Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Plexin D1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems AF4160
This item is not returnable.
View return policy
Description
Plexin D1 Polyclonal specifically detects Plexin D1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.Specifications
Plexin D1 | |
Polyclonal | |
Unconjugated | |
Q9Y4D7 | |
PLXND1 | |
Mouse myeloma cell line NS0-derived recombinant human Plexin D1 Leu47-Ala1271 Accession # Q9Y4D7 | |
100 μg | |
Primary | |
Detects human Plexin D1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Plexin A1, rmPlexin A3, and recombinant human Plexin B1 is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
LYOPH | |
Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-15 ug/mL, Neutralization 0.5-2 ug/mL, CyTOF-ready | |
KIAA0620MGC75353, PLEXD1, plexin D1, plexin-D1, PLXND1 | |
Goat | |
Affinity Purified | |
RUO | |
23129 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction