Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PlGF-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7758SP
This item is not returnable.
View return policy
Description
PlGF-3 Monoclonal antibody specifically detects PlGF-3 in Human samples. It is validated for ImmunohistochemistrySpecifications
PlGF-3 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5281 | |
Human | |
Purified |
Immunohistochemistry | |
1039018 | |
Immunohistochemistry 5-25 ug/mL | |
P49763 | |
Spodoptera frugiperda Sf21 (baculovirus)-derived human PlGF-3, Leu19-Arg221, Accession # P49763 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction