Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PlGF-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $409.00
Specifications
Antigen | PlGF-4 |
---|---|
Clone | 1039228 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2644SP
![]() |
R&D Systems
MAB2644SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2644100
![]() |
R&D Systems
MAB2644100 |
100 μg |
Each for $409.00
|
|
|||||
Description
PlGF-4 Monoclonal antibody specifically detects PlGF-4 in Human samples. It is validated for ImmunocytochemistrySpecifications
PlGF-4 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
5228 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
1039228 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P49763-4 | |
Human embryonic kidney cell HEK293-derived human PlGF-4, Leu19-Arg242, Accession # P49763-4 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title