Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PlGF Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $526.50
Specifications
Antigen | PLGF |
---|---|
Clone | 1038928 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11199SP
![]() |
R&D Systems
MAB11199SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11199100
![]() |
R&D Systems
MAB11199100 |
100 μg |
Each for $526.50
|
|
|||||
Description
PLGF Monoclonal antibody specifically detects PLGF in Human samples. It is validated for nanSpecifications
PLGF | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
D12S1900, PGF, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
CHO-derived human PlGF-2 protein,, Leu19-Arg170, Accession # NP_002623 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1038928 | |
Unconjugated | |
Mouse | |
Human | |
NP_002623 | |
5228 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title